accessibility menu, dialog, popup

UserName

FFFOHNNNN

CAS RN 370-86-5

IUPAC Name: 1-cyano-N-[4-(trifluoromethoxy)phenyl]methanecarbohydrazonoyl cyanide
Synonyms: 1-cyano-N-[4-(trifluoromethoxy)phenyl]methanecarbohydrazonoyl cyanide
Molecular Weight (g/mol): 254.17
Molecular Formula: C10H5F3N4O
InChi Key: BMZRVOVNUMQTIN-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(NN=C(C#N)C#N)C=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results