Learn More
OHAUS Carbon Steel Defender 3000 Series Floor Scale
Defender 3000 Floor Platforms meet dry weighing needs for industrial and commercial use. Their high-quality construction is designed to minimize flexing under heavy loads for years of dependable performance. The indicator features a bright display that changes color when in check mode. Scale also includes counting and totalization modes.
Supplier:  OHAUS 30837604
 View more versions of this product
Catalog No. 01-920-191
Only null left
Explore available promotions
Description
Description
  • Painted carbon steel treaded decks with four alloy steel load cells, fixed top plate (5 mm/0.196 in. thick) with structural bracing for stiff construction.
  • Platforms available in sizes of 3 x 3 ft. , 4 x 4 ft. , and 5 x 5 ft. and capacities from 2,500 lb. to 5,000 lb
  • Results are clearly shown on the large, bright backlit LCD display.
  • In checkweighing mode, display changes color (yellow/green/red) to alert users when within the target weight range.
Specifications
Specifications
2500 kg
7-segment Multi-color Backlit LCD
Class III
Scale
0.5 kg
i-DF33P5000B1L
Product Suggestions
Product Suggestions