missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Standard, (Potassium Hydrogen Phthalate), Certified, 1000ppm ±10ppm Organic (1mL = 1mg C) , LabChem™
Supplier: LabChem LC129101
Specifications
| Carbon Standard | |
| Poly Bottle | |
| 99.49,0.3,0.21 | |
| Liquid | |
| 1000 ppm ±10 ppm Organic (1mL = 1mg C) | |
| Colorless | |
| IWZKICVEHNUQTL-UHFFFAOYSA-M | |
| potassium;2-carboxybenzoate | |
| 500 mL | |
| 204.23 |
| 1000ppm, Organic (1mL = 1mg C) (Potassium Hydrogen Phthalate) | |
| 877-24-7 , 7664-38-2 , 877-24-7 , 7732-18-5 | |
| Passes Test | |
| C8H5KO4 | |
| KHC8H4O4 | |
| Soluble in water | |
| C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] | |
| 204.222 | |
| 23676735 | |
| Certified |
Chemical Identifiers
| 877-24-7 | |
| 204.222 | |
| 23676735 | |
| C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] |
| C8H5KO4 | |
| IWZKICVEHNUQTL-UHFFFAOYSA-M | |
| potassium;2-carboxybenzoate |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature