missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Standard, Organic, 2000 ppm, Reagents
Carbon Standard, Organic, 2000 ppm, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.
$307.07 - $307.07
Specifications
| Chemical Name or Material | Carbon Standard, Organic, 2000 ppm |
|---|---|
| CAS | 7732-18-5,877-24-7 |
| Specific Gravity | 1.002 |
| Physical Form | Liquid |
| Color | Colorless |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
01-335-590
|
Labchem Inc
CC2056001A |
1 L | Poly Bottle |
Each for $307.07
|
|
||||
|
01-335-589
|
Labchem Inc
CC2056001C |
1 L | Glass Bottle |
Each for $307.07
|
|
||||
Specifications
| Carbon Standard, Organic, 2000 ppm | |
| 1.002 | |
| Colorless | |
| Water Analyzer | |
| Water Analysis | |
| Reagent |
| 7732-18-5,877-24-7 | |
| Liquid | |
| 100°C | |
| Completely miscible | |
| 1 L |