missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Standard, Organic, 2000 ppm, Reagents
Carbon Standard, Organic, 2000 ppm, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CC2056001C
Specifications
| Carbon Standard, Organic, 2000 ppm | |
| 7732-18-5,877-24-7 | |
| Liquid | |
| 100°C | |
| Completely miscible | |
| 1 L |
| Glass Bottle | |
| 1.002 | |
| Colorless | |
| Water Analyzer | |
| Water Analysis | |
| Reagent |