missing translation for 'onlineSavingsMsg'
Learn More

Organic Carbon Standard, 1000 ppm C, Ricca Chemical

$101.30 - $1599.27

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
RC18474
View Documents
Ricca Chemical Company
18474
120 mL Amber Glass Bottle
Each for $101.30
Only null left
 
1847-16
SDS View Documents
Ricca Chemical Company
184716
500 mL Amber Glass Bottle
Each for $221.53
Only null left
 
1847-32
View Documents
Ricca Chemical Company
184732
1 L Amber Glass Bottle
Each for $267.73
Only null left
 
1847-25
View Documents
Ricca Chemical Company
184725
10 L Cubitainer
Each for $1,599.27
Only null left
 
Description

Description

Specifications

Chemical Identifiers

877-24-7
204.222
23676735
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
C8H5KO4
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate

Specifications

Amber Glass Bottle
1000ppm
100.0,0.48,0.22
Liquid
Colorless
100°C
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
204.222
23676735
877-24-7 , 7664-38-2 , 877-24-7 , 7732-18-5
97.0,0.46,0.21
1 mL = 1 mg C
C8H5KO4
< 2
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
120 mL
Organic Carbon Standard
Videos
SDS
Documents

Documents