Learn More
Carbon disulfide, 99.9%, for spectroscopy
CAS: 75-15-0 | CS2 | 76.131 g/mol
Supplier: Thermo Scientific Chemicals 167710025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopySpecifications
| Carbon disulfide | |
| 75-15-0 | |
| 1.2620g/mL | |
| 2.5 L | |
| Glass Bottle | |
| 1.627 | |
| 99.9% | |
| CS2 | |
| MFCD00011321 | |
| 0.03% max. | |
| 01,114; 05,94; 06,95 | |
| carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide | |
| QGJOPFRUJISHPQ-UHFFFAOYSA-N | |
| methanedithione | |
| 6348 | |
| 76.13 | |
| Spectroscopy |
| -111.0°C | |
| 0.01 max. at 420nm, 0.05 max. at 410nm, 0.1 max. at 400nm, 0.5 max. at 390nm, 1.3 max. at 380nm | |
| 46.0°C | |
| −30°C | |
| Liquid | |
| 0.0005% max. | |
| 1.262 | |
| CS2 | |
| 0.363 mPa.s (20°C) | |
| 03, 197 | |
| 15, 1811 | |
| Solubility in water: 2.9g/L (20°C). Other solubilities: miscible with anhydrous methanol, ethanol, ether,, benzene, chloroform, carbon tetrachloride and oils | |
| C(=S)=S | |
| 76.131 | |
| CHEBI:23012 | |
| 99.9% |
Chemical Identifiers
| 75-15-0 | |
| 76.131 | |
| QGJOPFRUJISHPQ-UHFFFAOYSA-N | |
| 6348 | |
| methanedithione |
| CS2 | |
| MFCD00011321 | |
| carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide | |
| CHEBI:23012 | |
| C(=S)=S |
Safety and Handling
GHS H Statement
Causes skin irritation. Causes serious eye irritation. Causes damage to organs through prolonged or repeated exposure. Suspected of damaging fertility. Suspected of damaging the unborn child. Highly flammable liquid and vapor. Harmful if inhaled.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
Danger
EINECSNumber : 200-843-6
RTECSNumber : FF6650000
TSCA : TSCA
RUO – Research Use Only