missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon disulfide, 99.9%, for spectroscopy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
Specifications
Specifications
| Melting Point | -111.0°C |
| Absorbance | 0.01 max. at 420nm, 0.05 max. at 410nm, 0.1 max. at 400nm, 0.5 max. at 390nm, 1.3 max. at 380nm |
| Density | 1.2620g/mL |
| Boiling Point | 46.0°C |
| Quantity | 25 mL |
| Flash Point | −30°C |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Refractive Index | 1.627 |
| Residue after Evaporation | 0.0005% max. |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. Causes damage to organs through prolonged or repeated exposure. Suspected of damaging fertility. Suspected of damaging the unborn child. Highly flammable liquid and vapor. Harmful if inhaled. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection. |
| EINECSNumber | 200-843-6 |
| RTECSNumber | FF6650000 |
| TSCA | TSCA |
RUO – Research Use Only