missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbimazole 95.0+%, TCI America™
$109.98 - $328.08
Chemical Identifiers
| CAS | 22232-54-8 |
|---|---|
| Molecular Formula | C7H10N2O2S |
| Molecular Weight (g/mol) | 186.229 |
| MDL Number | MFCD00027421 |
| InChI Key | CFOYWRHIYXMDOT-UHFFFAOYSA-N |
| Synonym | carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat |
| PubChem CID | 31072 |
| ChEBI | CHEBI:617099 |
| IUPAC Name | ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate |
| SMILES | CCOC(=O)N1C=CN(C1=S)C |
Chemical Identifiers
| 22232-54-8 | |
| 186.229 | |
| CFOYWRHIYXMDOT-UHFFFAOYSA-N | |
| 31072 | |
| ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate |
| C7H10N2O2S | |
| MFCD00027421 | |
| carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat | |
| CHEBI:617099 | |
| CCOC(=O)N1C=CN(C1=S)C |
Specifications
| 22232-54-8 | |
| White-Yellow | |
| MFCD00027421 | |
| carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat | |
| CCOC(=O)N1C=CN(C1=S)C | |
| 186.229 | |
| CHEBI:617099 | |
| ≥95.0% (HPLC) | |
| Carbimazole |
| 124°C | |
| C7H10N2O2S | |
| 5 g | |
| CFOYWRHIYXMDOT-UHFFFAOYSA-N | |
| ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate | |
| 31072 | |
| 186.23 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : NJ2441000
TSCA : No