accessibility menu, dialog, popup

UserName

CAS RN 22232-54-8

IUPAC Name: ethyl 3-methyl-2-sulfanylidene-2,3-dihydro-1H-imidazole-1-carboxylate
Synonyms: carbimazole neo-thyreostat herbrand brand of carbimazole carbotiroid carbimazol atirozidina neo-tireol thyrostat carbimazolum carbimazol henning
Molecular Weight (g/mol): 186.23
Molecular Formula: C7H10N2O2S
InChi Key: CFOYWRHIYXMDOT-UHFFFAOYSA-N
SMILES: CCOC(=O)N1C=CN(C)C1=S
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)