missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Carbimazole | |
| 124°C | |
| C7H10N2O2S | |
| 25 g | |
| CFOYWRHIYXMDOT-UHFFFAOYSA-N | |
| ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate | |
| 31072 | |
| 186.23 | |
| Crystalline Powder |
| 22232-54-8 | |
| White-Yellow | |
| MFCD00027421 | |
| carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat | |
| CCOC(=O)N1C=CN(C1=S)C | |
| 186.229 | |
| CHEBI:617099 | |
| ≥95.0% (HPLC) |
Chemical Identifiers
| 22232-54-8 | |
| 186.229 | |
| CFOYWRHIYXMDOT-UHFFFAOYSA-N | |
| 31072 | |
| ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate |
| C7H10N2O2S | |
| MFCD00027421 | |
| carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat | |
| CHEBI:617099 | |
| CCOC(=O)N1C=CN(C1=S)C |
Safety and Handling
RTECSNumber : NJ2441000
TSCA : No