missing translation for 'onlineSavingsMsg'
Learn More

Carbazole-9-carbonyl Chloride 98.0+%, TCI America™

$202.15 - $1543.98

Chemical Identifiers

CAS 73500-82-0
Molecular Formula C13H8ClNO
Molecular Weight (g/mol) 229.663
MDL Number MFCD00059689
InChI Key RCENRFKQKSIRLA-UHFFFAOYSA-N
Synonym 9-(Chlorocarbonyl)carbazole
PubChem CID 3262261
IUPAC Name carbazole-9-carbonyl chloride
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C(=O)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C10311G
View Documents
TCI America
C10311G
1 g
Each for $202.15
Only null left
 
C10315G
View Documents
TCI America
C10315G
5 g
Each for $573.70
Only null left
 
C103125G
View Documents
TCI America
C103125G
25 g
Each for $1,543.98
Only null left
 
Description

Description

Specifications

Chemical Identifiers

73500-82-0
229.663
RCENRFKQKSIRLA-UHFFFAOYSA-N
3262261
C1=CC=C2C(=C1)C3=CC=CC=C3N2C(=O)Cl
C13H8ClNO
MFCD00059689
9-(Chlorocarbonyl)carbazole
carbazole-9-carbonyl chloride

Specifications

73500-82-0
White
MFCD00059689
9-(Chlorocarbonyl)carbazole
C1=CC=C2C(=C1)C3=CC=CC=C3N2C(=O)Cl
229.663
229.66
Crystalline Powder
105°C
C13H8ClNO
1 g
RCENRFKQKSIRLA-UHFFFAOYSA-N
carbazole-9-carbonyl chloride
3262261
≥98.0% (GC,T)
Carbazole-9-carbonyl Chloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents