accessibility menu, dialog, popup

UserName

NOCl

CAS RN 73500-82-0

IUPAC Name: 9H-carbazole-9-carbonyl chloride
Synonyms: carbazole-9-carbonyl chloride
Molecular Weight (g/mol): 229.66
Molecular Formula: C13H8ClNO
InChi Key: RCENRFKQKSIRLA-UHFFFAOYSA-N
SMILES: ClC(=O)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results