missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbazole-9-carbonyl Chloride 98.0+%, TCI America™
Supplier: TCI America C10315G
Specifications
| Carbazole-9-carbonyl Chloride | |
| 105°C | |
| C13H8ClNO | |
| 5 g | |
| RCENRFKQKSIRLA-UHFFFAOYSA-N | |
| carbazole-9-carbonyl chloride | |
| 3262261 | |
| ≥98.0% (GC,T) |
| 73500-82-0 | |
| White | |
| MFCD00059689 | |
| 9-(Chlorocarbonyl)carbazole | |
| C1=CC=C2C(=C1)C3=CC=CC=C3N2C(=O)Cl | |
| 229.663 | |
| 229.66 | |
| Crystalline Powder |
Chemical Identifiers
| 73500-82-0 | |
| 229.663 | |
| RCENRFKQKSIRLA-UHFFFAOYSA-N | |
| 3262261 | |
| C1=CC=C2C(=C1)C3=CC=CC=C3N2C(=O)Cl |
| C13H8ClNO | |
| MFCD00059689 | |
| 9-(Chlorocarbonyl)carbazole | |
| carbazole-9-carbonyl chloride |
Safety and Handling
TSCA : No