missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbazochrome 99.0+%, TCI America™
$41.02 - $109.98
Chemical Identifiers
| CAS | 69-81-8 |
|---|---|
| Molecular Formula | C10H12N4O3 |
| Molecular Weight (g/mol) | 236.231 |
| MDL Number | MFCD00059705 |
| InChI Key | XSXCZNVKFKNLPR-UHFFFAOYSA-N |
| Synonym | Adrenochrome Semicarbazone |
| PubChem CID | 5478929 |
| IUPAC Name | [(3-hydroxy-1-methyl-6-oxo-2,3-dihydroindol-5-ylidene)amino]urea |
| SMILES | CN1CC(C2=CC(=NNC(=O)N)C(=O)C=C21)O |
Chemical Identifiers
| 69-81-8 | |
| 236.231 | |
| XSXCZNVKFKNLPR-UHFFFAOYSA-N | |
| 5478929 | |
| CN1CC(C2=CC(=NNC(=O)N)C(=O)C=C21)O |
| C10H12N4O3 | |
| MFCD00059705 | |
| Adrenochrome Semicarbazone | |
| [(3-hydroxy-1-methyl-6-oxo-2,3-dihydroindol-5-ylidene)amino]urea |
Specifications
| 69-81-8 | |
| C10H12N4O3 | |
| 5 g | |
| XSXCZNVKFKNLPR-UHFFFAOYSA-N | |
| [(3-hydroxy-1-methyl-6-oxo-2,3-dihydroindol-5-ylidene)amino]urea | |
| 5478929 | |
| ≥99.0% (HPLC) | |
| Carbazochrome |
| 203°C | |
| MFCD00059705 | |
| Adrenochrome Semicarbazone | |
| CN1CC(C2=CC(=NNC(=O)N)C(=O)C=C21)O | |
| 236.231 | |
| 236.23 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : NM1925500
TSCA : No