missing translation for 'onlineSavingsMsg'
Learn More

Carbazochrome 99.0+%, TCI America™

Catalog No. a01765g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A01765G 5 g
A017625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A01765G Supplier TCI America Supplier No. A01765G
Only null left

Chemical Identifiers

CAS 69-81-8
Molecular Formula C10H12N4O3
Molecular Weight (g/mol) 236.231
MDL Number MFCD00059705
InChI Key XSXCZNVKFKNLPR-UHFFFAOYSA-N
Synonym Adrenochrome Semicarbazone
PubChem CID 5478929
IUPAC Name [(3-hydroxy-1-methyl-6-oxo-2,3-dihydroindol-5-ylidene)amino]urea
SMILES CN1CC(C2=CC(=NNC(=O)N)C(=O)C=C21)O

Specifications

Melting Point 203°C
Quantity 5 g
Formula Weight 236.23
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Carbazochrome
RTECSNumber NM1925500
TSCA No