accessibility menu, dialog, popup

UserName

H3CNHONNHNH2OO(Z)

CAS RN 69-81-8

IUPAC Name: {[(5Z)-3-hydroxy-1-methyl-6-oxo-2,3,5,6-tetrahydro-1H-indol-5-ylidene]amino}urea
Synonyms: carbazochrome 卡巴克络
Molecular Weight (g/mol): 236.23
Molecular Formula: C10H12N4O3
InChi Key: XSXCZNVKFKNLPR-SDQBBNPINA-N
SMILES: CN1CC(O)C2=C\C(=N\NC(N)=O)C(=O)C=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results