missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ IgA-CH1 (Hu) Affinity Matrix
GREENER_CHOICE

Catalog No. 194311005
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
194311005 5 mL
194311010 10 mL
194311050 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 194311005 Supplier Thermo Scientific™ Supplier No. 194311005

Thermo Scientific™ CaptureSelect™ IgA-CH1 (Hu) Affinity Matrix can be used to purify human IgA.

CaptureSelect IgA-CH1 (Hu) Affinity Matrix can be used to purify human IgA. It shows no binding to other species IgA or other immunoglobulins.

This product can be used for purification of:

  • IgA1 and IgA2 from plasma and serum
  • Mono- and dimeric IgA, including secretory IgA
  • IgA Fab fragments

Specifications

Content And Storage Store at 2°C to 8°C.
Form Liquid Suspension
Product Line CaptureSelect
Quantity 5 mL
Type Affinity Matrix
Stationary Phase Anti-IgA-CH1 (Hu)