missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Human Fab-lambda Kinetics Biotin Conjugate
GREENER_CHOICE

Catalog No. 7103312500
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7103312500 500 μg
7103312100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7103312500 Supplier Thermo Scientific™ Supplier No. 7103312500
Get an Extra Analytical Consumable with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ CaptureSelect™ Human Fab-lambda Kinetics Biotin Conjugate is a bi-functional ligand construct consisting of 2 different 13 kDa recombinant single domain antibody fragments (affinity ligands).

CaptureSelect Human Fab-lambda Kinetics Biotin Conjugate is a bi-functional ligand construct consisting of 2 different 13 kDa recombinant single domain antibody fragments (affinity ligands).

The CaptureSelect anti-LC-lambda (Hu) ligand that specifically binds to the constant domain of human lambda light chains is genetically fused to the CaptureSelect anti-IgG-CH1 ligand that specifically binds to the CH1 domain of all four human IgG subclasses. The bi-functional affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Human Fab-lambda Kinetics Biotin Conjugate allows you to:

  • Screen Fab-antigen interactions—the Human Fab-lambda Kinetics Biotin Conjugate allows high affinity capture of human IgG Fab fragments that possess a lambda light chain. Extreme low off-rates [k(diss)] are achieved through avidity by targeting the CL-lambda domain of the Fab (light chain) and its IgG CH1 domain (heavy chain) simultaneously, thereby enabling Fab screening on target binding kinetics.

Applications for CaptureSelect Human Fab-lambda Kinetics Biotin Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Form Liquid
Product Line CaptureSelect
Quantity 500 μg
Type Conjugate
Label or Dye Biotin & Other Haptens
Ligand Type Antibody