Learn More
Thermo Scientific™ CaptureSelect™ FSH Affinity Matrix
Thermo Scientific™ CaptureSelect™ FSH Affinity Matrix has been specifically designed for the purification of follicle stimulating hormone (FSH) from recombinant sources.
$100639.23 - $100639.23
Specifications
| Flow Rate | 200 to 400 cm/hr. |
|---|---|
| Matrix | Agarose-based, aldehyde-activated |
| Particle Size | 65 μm |
| Form | Liquid Suspension |
| Product Line | CaptureSelect |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
19-431-801L
|
Thermo Scientific™
19431801L |
1 L |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
CaptureSelect FSH Affinity Matrix has been specifically designed for the purification of follicle stimulating hormone (FSH) from recombinant sources.
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Specifications
| 200 to 400 cm/hr. | |
| 65 μm | |
| CaptureSelect | |
| Anti-Follicle Stimulating Hormone |
| Agarose-based, aldehyde-activated | |
| Liquid Suspension | |
| Affinity Matrix |