missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ FcXL Affinity Matrix

Catalog No. 501147824
Encompass
Change view
Click to view available options
Quantity:
5 mL
50 mL
250 mL
1 L
5 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501147824 5 mL
501147826 50 mL
361054481 250 mL
361054482 1 L
361054483 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 501147824 Supplier Thermo Scientific™ Supplier No. 194328005

Thermo Scientific™ CaptureSelect™ FcXL Affinity Matrix has been specifically designed for the purification of recombinant human IgG, Fc fusion proteins, and plasma derived IgG.

CaptureSelect FcXL Affinity Matrix has been specifically designed for the purification of recombinant human IgG, Fc fusion proteins, and plasma derived IgG. It recognizes the CH3 domain of all human IgG subclasses (IgG1, 2, 3, and 4).

Features of this affinity matrix include:

  • Mild elution, making it suitable for Fc fusion proteins
  • Human specific, no binding to bovine antibodies, for example
  • Excellent scalability
  • Non-animal-derived

CaptureSelect FcXL Affinity Matrix purifies IgGs and Fc fusion proteins directly from complex source materials in a single step with high purity and yield.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main Characteristics:

  • Matrix: agarose-based, aldehyde-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect FcXL affinity ligand
  • Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
  • Binding capacity: 25–35 g IgG/L resin depending on flow rate, column height, and contact time
  • Elution conditions: 20 mM citric acid or acetic acid, pH 3–4, or 20 mM sodium acetate, 1.0 M MgCl2, 40% (v/v) propylene glycol, pH 5–6
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20%(v/v) ethanol

Specifications

Binding Capacity 25 to 35 g IgG/L
Flow Rate 50 to 200 cm/hr. (Up to 2 bar)
Matrix Agarose-based, aldehyde-activated
Content And Storage Store in refrigerator (2°C to 8°C).
Particle Size 65 ±10 μm
Form Agarose Bead
Product Line CaptureSelect
Quantity 5 mL
For Use With (Application) IgG purification
Target Fc of IgG, CH3 domain
Type Affinity Matrix
Ligand Type FcXL affinity ligand
Stationary Phase Anti-IgG-Fc
Show More Show Less