Learn More
Thermo Scientific™ CaptureSelect™ CH1-XL RoboColumn™ Pre-packed Column
Thermo Scientific™ CaptureSelect™ RoboColumn™ small chromatography columns are used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.
$2685.54 - $2685.54
Specifications
| Binding Capacity | >15 mg/mL human Fab fragment |
|---|---|
| Flow Rate | 16 to 150 cm/hr. |
| Matrix | Agarose-based, epoxide-activated |
| Content And Storage | Store in refrigerator (2°C to 8°C) |
| Diameter (Metric) | 5 mm |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|
59-434-62600
|
Thermo Scientific™
5943462600 |
600 μL |
Each for $2,685.54
|
|
|||||
Description
CaptureSelect RoboColumn™ small chromatography columns are used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.
CaptureSelect CH1-XL Affinity Matrix recognizes the CH1 domain of human IgG antibodies, thereby enabling purification of all Fab fragments, independent of the light-chain type (kappa or lambda) and without binding of over-expressed light chain and light-chain dimers. In addition, it can be used for the purification of human IgGs from all four subclasses (1, 2, 3, and 4).
Features of this affinity matrix include:
- Platform approach for purification of high quality and correctly assembled human Fab fragments
- No co-purification of over-expressed free light chains due to selectivity for the CH1 domain
- Elution possible at pH 4.0–4.5 with 50 mM acetate buffer
- Caustic cleaning with up to 50 mM NaOH
- Excellent scalability
- No cross-binding to bovine IgG, therefore presence of fetal bovine serum is not a problem
- Non-animal-derived
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Characteristics
- Column size: 200 μL and 600 μL, 1 strip, 8 columns (8-row)
- Column dimension: 200 μL: 5 x 10 mm
- Column dimension: 600 μL: 5 x 30 mm
- Matrix: agarose based, epoxide-activated
- Average particle size: 65 ± 10 μm
- Ligand: CaptureSelect CH1-XL affinity ligand
- Ligand coupling: epoxide coupling
- Binding capacity: >15 mg/mL human Fab fragment depending on flow rate, column height, contact time. For humanized Fabs, lower capacities may be observed.
- Elution conditions: 100 mM glycine buffer, pH 3.0; 50 mM acetate buffer, pH 4.0–4.5
- Formulation buffer: 20% (v/v) ethanol
- Operating pressure: 2 bar (maximum)
- Flow rate: 16–150 cm/h
RoboColumn is a trademark of Repligen.
Specifications
| >15 mg/mL human Fab fragment | |
| Agarose-based, epoxide-activated | |
| 5 mm | |
| Room temperature, or on wet ice | |
| CaptureSelect | |
| Column | |
| CH1-XL affinity ligand on agarose bead | |
| RoboColumn |
| 16 to 150 cm/hr. | |
| Store in refrigerator (2°C to 8°C) | |
| 2 bar | |
| 65 ±10 μm | |
| Chromatography | |
| CH1-XL affinity ligand | |
| 8-well Strips |