missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ CH1-XL Magnetic Agarose Beads
GREENER_CHOICE

Catalog No. 2883462050
Change view
Click to view available options
Quantity:
1 mL
5 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2883462050 50 mL
2883462001 1 mL
2883462005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 2883462050 Supplier Thermo Scientific™ Supplier No. 2883462050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ CaptureSelect™ CH1-XL Magnetic Agarose Beads are high-capacity, high-throughput magnetic affinity particles for antibody purification using manual or robotic magnetic separators.

CaptureSelect CH1-XL Magnetic Agarose Beads are high-capacity, high-throughput magnetic affinity particles for antibody purification using manual or robotic magnetic separators. The CaptureSelect CH1-XL affinity ligand recognizes the CH1 domain of human IgG, enabling purification of all subclasses (1, 2, 3, and 4) of human IgG and Fab fragments, independent of the light-chain isotype and source material. Due to selectivity for the CH1 domain, no co-purification of free light chains will occur. IgG and Fab fragments can be directly isolated from complex source materials in a single step with high purity and yield.

Features of CaptureSelect CH1-XL Magnetic Agarose Beads include:

  • One-step human IgG purification with high purity, from crude material
  • Compatible with manual and automated workflows (e.g., Thermo Scientific KingFisher instruments)
  • Short operation time
  • Mild elution at pH 4
  • No sample dilution required during isolation, repetitive sample loading feasible
  • No binding to bovine antibodies, allowing use of fetal bovine serum in the sample material
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: 6% cross-linked magnetized agarose
  • Average particle size: 25–30 μm
  • Suspension: 25% slurry
  • Ligand: CaptureSelect CH1-XL affinity ligand
  • Binding capacity: >4 mg IgG/mL slurry
  • Elution conditions: 50 mM sodium acetate, pH 4.0/0.1 M glycine, pH 3.0

Specifications

Binding Capacity >4 mg IgG/mL slurry
Matrix 6% cross-linked magnetized agarose
Particle Size 25 to 30 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 50 mL
Target CH1 domain of human IgG
Type Magnetic Agarose Bead
Ligand Type CH1-XL affinity ligand