Learn More
Thermo Scientific™ CaptureSelect™ CH1-XL Affinity Matrix

Description
CaptureSelect CH1-XL Affinity Matrix is the improved version of CaptureSelect IgG-CH1 Affinity Matrix. The specificity is the same, but it features a wider elution range (pH 3.0-4.5), increased binding capacity, and improved stability at higher pH.
CaptureSelect CH1-XL Affinity Matrix recognizes the CH1 domain of human IgG antibodies, enabling purification of all subclasses (1, 2, 3, and 4) of human IgG and Fab fragments, independent of the light-chain isotype and source material.
CaptureSelect CH1-XL Affinity Matrix features include:
- Platform approach for purification of IgGs and their Fab fragments directly from complex source materials in a single step with high purity and yield
- Elution possible at pH 4.0–4.5 with 50 mM acetate buffer
- No co-purification of overproduced free light chain due to selectivity for the CH1 domain
- No binding to bovine antibodies, so presence of fetal bovine serum is not a problem
- Excellent scalability
- Non-animal derived
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Main characteristics:
- Matrix: agarose-based, epoxide-activated
- Average particle size: 65 ± 10 μm
- Ligand: CaptureSelect CH1-XL affinity ligand
- Ligand coupling method: epoxide coupling
- Binding capacity: >15 mg/mL human Fab fragment depending on flow rate, column height, contact time. For humanized Fabs lower capacities can be observed.
- Elution conditions: 100 mM glycine buffer, pH 3.0; 50 mM acetate buffer, pH 4.0–4.5
- Flow rate: 50–200 cm/h (up to 2 bar)
- Formulation buffer: 20% (v/v) ethanol
Specifications
Specifications
| Content And Storage | Store at 2°C to 8°C. |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice, Wet Ice |
| Stationary Phase | Anti-IgG-CH1 |
| Product Line | CaptureSelect |
| Type | Affinity Matrix |
| Quantity | 1 L |
| Form | Agarose Bead |
| Particle Size | 65 μm |
| Binding Capacity | >15 mg/mL |
| Flow Rate | 50 to 200 cm/hr. (Up to 2 bar) |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.