missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-IgM Conjugate
GREENER_CHOICE

Catalog No. p-7180820
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7102892100 100 μg
7102892500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7102892100 Supplier Thermo Scientific™ Supplier No. 7102892100
Only null left

Thermo Scientific™ CaptureSelect™ Biotin Anti-IgM Conjugate consists of a 14 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the μ chain of human, rat, and mouse IgM antibodies.

CaptureSelect Biotin Anti-IgM Conjugate consists of a 14 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the μ chain of human, rat, and mouse IgM antibodies. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Biotin Anti-IgM Conjugate allows you to:

  • Detect, quantitate, and characterize all human, rat, and mouse IgM antibodies (independent of the type of light chain), recombinant IgM monoclonal antibodies, and total IgM in human plasma and/or serum samples
  • Avoid cross-binding with other isotypes such as IgG, IgA, IgD, and IgE, free light chains, or with IgM from bovine sources such as FCS

Applications for CaptureSelect Biotin Anti-IgM Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Content And Storage Store at -5°C to -30°C.
Form Liquid
Product Line CaptureSelect
Quantity 100 μg
Target IgM
Type Conjugate
Label or Dye Biotin & Other Haptens
Ligand Type Antibody