missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-IgA Conjugate
GREENER_CHOICE

Catalog No. 7102882100
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7102882100 100 μL
7102882500 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7102882100 Supplier Thermo Scientific™ Supplier No. 7102882100

Thermo Scientific™ CaptureSelect™ Biotin Anti-IgA Conjugate consists of a 14 kDa recombinant single domain antibody fragment that specifically binds to the α-chain of all human IgA antibodies (Fc domain).

CaptureSelect Biotin Anti-IgA Conjugate consists of a 14 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the α-chain of all human IgA antibodies (Fc domain). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Biotin Anti-IgA Conjugate allows you to:

  • Detect, quantitate, and characterize all human IgA antibodies, including monomeric IgA-1 and IgA-2, dimeric IgA (secretory and non-secretory), and recombinant IgA monoclonal antibodies, as well as total IgA in human plasma and/or serum samples
  • Avoid cross-binding with other isotypes, such as IgG, IgM, IgD, and IgE, free light chains, or IgA from bovine sources such as FCS

Applications for CaptureSelect Biotin Anti-IgA Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Content And Storage Store at -5°C to -30°C.
Form Liquid
Product Line CaptureSelect
Quantity 100 μL
Target IgA
Type Conjugate
Label or Dye Biotin & Other Haptens
Ligand Type Antibody