Learn More
Thermo Scientific™ CaptureSelect™ Biotin Anti-EPO Conjugate
Thermo Scientific™ CaptureSelect™ Biotin Anti-EPO Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant and natural-sourced human erythropoietin (EPO).
$1,536.04 - $5,409.96
Specifications
| Content And Storage | Store at -5°C to -30°C. |
|---|---|
| Label Type | Biotin and Other Haptens |
| Product Line | CaptureSelect |
| Product Type | Ligand Conjugates |
| Target Species | Human |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
71-033-72100
|
Thermo Scientific™
7103372100 |
100 μg |
N/A
|
|
|||||
|
71-033-72500
|
Thermo Scientific™
7103372500 |
500 μg |
N/A
|
|
|||||
Description
CaptureSelect Biotin Anti-EPO Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant and natural-sourced human erythropoietin (EPO). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.
Applications for CaptureSelect Biotin Anti-EPO Conjugate include ELISA, western blot, Gyrolab™-based immunoassays, and use of label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Specifications
| Store at -5°C to -30°C. | |
| CaptureSelect | |
| Human | |
| Approved for shipment on Wet or Dry Ice |
| Biotin and Other Haptens | |
| Ligand Conjugates | |
| Antibody |