missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-EPO Conjugate
GREENER_CHOICE

Catalog No. 7103372500
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7103372500 500 μg
7103372100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7103372500 Supplier Thermo Scientific™ Supplier No. 7103372500
Only null left

Thermo Scientific™ CaptureSelect™ Biotin Anti-EPO Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant and natural-sourced human erythropoietin (EPO).

CaptureSelect Biotin Anti-EPO Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant and natural-sourced human erythropoietin (EPO). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

Applications for CaptureSelect Biotin Anti-EPO Conjugate include ELISA, western blot, Gyrolab™-based immunoassays, and use of label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Content And Storage Store at -5°C to -30°C.
Label Type Biotin and Other Haptens
Product Line CaptureSelect
Product Type Ligand Conjugates
Target Species Human
Ligand Type Antibody
Quantity 500 μg
Shipping Condition Approved for shipment on Wet or Dry Ice