missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-CD63 Conjugate
GREENER_CHOICE

Catalog No. 7103992100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7103992100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 7103992100 Supplier Thermo Scientific™ Supplier No. 7103992100

Thermo Scientific™ CaptureSelect™ Biotin Anti-CD63 Conjugate consists of a 12–15 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to the tetraspanin marker CD63.

CaptureSelect Biotin Anti-CD63 Conjugate consists of a 12–15 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to the tetraspanin marker CD63. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

This conjugate allows you to detect, quantitate, and characterize CD63-expressing extracellular vesicles (EVs) like exosomes. Together with CaptureSelect HRP Anti-CD63 Conjugate (Cat. No. 7303992100), a sandwich ELISA can be set up to detect and quantitate CD63 expressing EVs.

Applications for CaptureSelect Biotin Anti-CD63 Conjugate include sandwich ELISA, Gyrolab™-based immunoassays, immunoprecipitation, and sample prep purification using streptavidin-coupled Dynabeads or MSIA Streptavidin D.A.R.T.S and label-free detection platforms such as those based on surface plasmon resonance (SPR) and bio-layer interferometry (BLI). For more information see our Biotin Application Guide.

For detection of a broad variety of EVs and exosomes that originate from different cell types and secretion pathways, such as those of endosomal (CD63-enriched, CD9+, or CD81+) or ectosomal (CD63 low, CD9+ and CD81+) origin, we also offer CaptureSelect Biotin and HRP anti-CD81 conjugates and CaptureSelect Biotin and HRP anti-CD9 conjugates.

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Concentration 1 mg/mL
Formulation Phosphate-buffered saline (pH 7.4)
Content And Storage Store in freezer (–5 to –30°C).
Shipping Condition Dry Ice
Reactivity Anti-tetraspanin CD63
Form Frozen Liquid
Product Line CaptureSelect
Quantity 100 μg
Target Tetraspanin CD63
Final Product Type Biotin Conjugate
Label or Dye Biotin
Ligand Type Recombinant single-domain antibody fragment (VHH ligand)
Show More Show Less