missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-AdV5 Conjugate
GREENER_CHOICE

Catalog No. 7103532100
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7103532100 100 μg
7103532500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7103532100 Supplier Thermo Scientific™ Supplier No. 7103532100

Thermo Scientific™ CaptureSelect™ Biotin Anti-AdV5 Conjugate consists of a 13-kDa recombinant single domain antibody fragment that specifically binds with high affinity and selectivity to hexon capsomeres of Adenovirus serotype 5 (AdV5) particles as well as free hexon proteins thereof.

CaptureSelect Biotin Anti-AdV5 Conjugate consists of a 13-kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to hexon capsomeres of Adenovirus serotype 5 (AdV5) particles as well as free hexon proteins thereof. Possible cross-reactivity towards hexon proteins of other Adenovirus serotypes has not been tested.

The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces. This conjugate allows you to detect, quantitate, and characterize AdV5 virus particles.

Applications for CaptureSelect Biotin Anti-AdV5 Conjugate include ELISA, immunoprecipitation with streptavidin-coupled Dynabeads, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Shipping Condition Dry Ice
Form Liquid
Product Line CaptureSelect
Quantity 100 μg
Type Conjugate
Label or Dye Biotin & Analogs