Learn More
Thermo Scientific™ CaptureSelect™ Biotin Anti-AAT Conjugate
Thermo Scientific™ CaptureSelect™ Biotin Anti-AAT Conjugate consists of a 13 kDa recombinant single domain antibody fragment that specifically binds with high affinity and selectivity to human alpha-1 antitrypsin (AAT).
$1514.85 - $5335.33
Specifications
| Content And Storage | Store in freezer (-5°C to -30°C). |
|---|---|
| Label Type | Biotin and Analogs |
| Product Line | CaptureSelect |
| Target Species | Human |
| Ligand Type | Antibody |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
71-028-72100
|
Thermo Scientific™
7102872100 |
100 μg |
Each for $1,514.85
|
|
|||||
|
71-028-72500
|
Thermo Scientific™
7102872500 |
500 μg |
Each for $5,335.33
|
|
|||||
Description
CaptureSelect Biotin Anti-AAT Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to human alpha-1 antitrypsin (AAT). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces. This conjugate allows you to detect, quantitate, and characterize native and recombinant human AAT.
Applications for CaptureSelect Biotin Anti-AAT Conjugate include ELISA, immunoprecipitation with streptavidin-coupled DynaBeads, Gyrolab™-based immunoassays, and label-free detection using platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Specifications
| Store in freezer (-5°C to -30°C). | |
| CaptureSelect | |
| Antibody |
| Biotin and Analogs | |
| Human |