Learn More
Captopril
Orally active inhibitor of angiotensin-converting enzyme | CAS: 62571-86-2 | C9H15NO3S | 217.283 g/mol
Supplier: Thermo Scientific Chemicals J6359314
Description
Captopril has also been shown to inhibit the formation of angiotensin II, a bioactive peptide that stimulates angiogenesis and increases microvessel density. Captopril demonstrates noncompetitive inhibition of tyrosinase monophenolase activity and competitive inhibition of diphenolase activity
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCaptopril has also been shown to inhibit the formation of angiotensin II, a bioactive peptide that stimulates angiogenesis and increases microvessel density. Captopril demonstrates noncompetitive inhibition of tyrosinase monophenolase activity and competitive inhibition of diphenolase activity
Solubility
Soluble in water (160 mg/ml) at 25°C, sesame and corn oils (<1mg/mL) at 25°C, methanol (>100 mg/ml), ethanol (>100 mg/ml), isopropanol (>100 mg/ml), chloroform (>100 mg/ml), and dichloromethane (>100 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
| Captopril | |
| 104°C to 108°C | |
| C9H15NO3S | |
| 25 g | |
| 14,1774 | |
| Soluble in water (160mg/ml) at 25°C,sesame and corn oils (<1mg/ml) at 25°C,methanol (>100mg/ml),ethanol (>100mg/ml),isopropanol (>100mg/ml),chloroform (>100mg/ml),and dichloromethane (>100mg/ml). | |
| CC(CS)C(=O)N1CCCC1C(=O)O | |
| 217.283 | |
| CHEBI:3380 | |
| Powder |
| 62571-86-2 | |
| White | |
| MFCD00168073 | |
| 477887 | |
| captopril, capoten, l-captopril, captopryl, lopirin, cesplon, captolane, tensoprel, acepress, captoril | |
| FAKRSMQSSFJEIM-RQJHMYQMSA-N | |
| (2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid | |
| 44093 | |
| 217.29 |
Chemical Identifiers
| 62571-86-2 | |
| 217.283 | |
| FAKRSMQSSFJEIM-RQJHMYQMSA-N | |
| 44093 | |
| (2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid |
| C9H15NO3S | |
| MFCD00168073 | |
| captopril, capoten, l-captopril, captopryl, lopirin, cesplon, captolane, tensoprel, acepress, captoril | |
| CHEBI:3380 | |
| CC(CS)C(=O)N1CCCC1C(=O)O |
Safety and Handling
GHS H Statement
H361-H317
Suspected of damaging fertility or the unborn child.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P305+P351+P338-P308+P313-P310-P333+P313-P363-P501c
H317-H318-H360D
EINECSNumber : 263-607-1
RTECSNumber : UY0550000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only