missing translation for 'onlineSavingsMsg'
Learn More

Captopril

Catalog No. p-7042740
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6359303 1 g
AAJ6359306 5 g
AAJ6359314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6359303 Supplier Thermo Scientific Chemicals Supplier No. J6359303

Orally active inhibitor of angiotensin-converting enzyme | CAS: 62571-86-2 | C9H15NO3S | 217.283 g/mol

Captopril has also been shown to inhibit the formation of angiotensin II, a bioactive peptide that stimulates angiogenesis and increases microvessel density. Captopril demonstrates noncompetitive inhibition of tyrosinase monophenolase activity and competitive inhibition of diphenolase activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Captopril has also been shown to inhibit the formation of angiotensin II, a bioactive peptide that stimulates angiogenesis and increases microvessel density. Captopril demonstrates noncompetitive inhibition of tyrosinase monophenolase activity and competitive inhibition of diphenolase activity

Solubility
Soluble in water (160 mg/ml) at 25°C, sesame and corn oils (<1mg/mL) at 25°C, methanol (>100 mg/ml), ethanol (>100 mg/ml), isopropanol (>100 mg/ml), chloroform (>100 mg/ml), and dichloromethane (>100 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 62571-86-2
Molecular Formula C9H15NO3S
Molecular Weight (g/mol) 217.283
MDL Number MFCD00168073
InChI Key FAKRSMQSSFJEIM-RQJHMYQMSA-N
Synonym captopril, capoten, l-captopril, captopryl, lopirin, cesplon, captolane, tensoprel, acepress, captoril
PubChem CID 44093
ChEBI CHEBI:3380
IUPAC Name (2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid
SMILES CC(CS)C(=O)N1CCCC1C(=O)O

Specifications

Melting Point 104°C to 108°C
Color White
Quantity 1 g
Beilstein 477887
Merck Index 14,1774
Solubility Information Soluble in water (160mg/ml) at 25°C,sesame and corn oils (<1mg/ml) at 25°C,methanol (>100mg/ml),ethanol (>100mg/ml),isopropanol (>100mg/ml),chloroform (>100mg/ml),and dichloromethane (>100mg/ml).
Formula Weight 217.29
Physical Form Powder
Chemical Name or Material Captopril
Hazard Category H317-H318-H360D
Hazard Statement GHS H Statement
H361-H317
Suspected of damaging fertility or the unborn child.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P305+P351+P338-P308+P313-P310-P333+P313-P363-P501c
EINECSNumber 263-607-1
RTECSNumber UY0550000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only