accessibility menu, dialog, popup

UserName

CAS RN 62571-86-2

IUPAC Name: (2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid
Synonyms: captopril tenosbon tensoprel garranil (discontinued) acepress lopril hipertil acepril aceplus apopril
Molecular Weight (g/mol): 217.28
Molecular Formula: C9H15NO3S
InChi Key: FAKRSMQSSFJEIM-RQJHMYQMSA-N
SMILES: C[C@H](CS)C(=O)N1CCC[C@H]1C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Physical Form 
  • (5)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (6)
  • (3)
  • (5)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)