missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Capto S™ ImpAct Chromatography Media
Description
- High-flow characteristics of Capto media allows for good pressure-flow properties
- High binding capacity and resolution, combined with the ability to run at high flow rates and bed heights, increases productivity and flexibility in process design
- Efficient aggregate removal at high load of monoclonal antibodies
Specifications
Specifications
| Product Type | Capto S ImpAct Chromatography Media |
| Quantity | 100 mL |
| Material | High-flow agarose |
| pH Range | 4 to 12 |
| Particle Size | 50μm |
| Matrix | Ion Exchange Chromatography Media |