missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto S™ ImpAct Chromatography Media

Catalog No. 45003200
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45003200 25 mL
45003201 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45003200 Supplier Cytiva Supplier No. 17371701
Only null left

Capto S ImpAct is a strong cation exchange chromatography medium (resin) developed for polishing of monoclonal antibodies (MAbs) and other biomolecules.

  • High binding capacity, > 100 mg MAb/mL medium.
  • Efficient aggregate removal at high load of monoclonal antibodies.
  • High-resolution polishing based on the well-established Capto platform.
  • Flexibility of design thanks to a large operational window of flow rates and bed heights for easy optimization and scaling.
  • High productivity enabling cost-efficient manufacturing.
  • Security of supply and comprehensive regulatory support.

Specifications

Product Type Capto S ImpAct Chromatography Media
Quantity 25 mL
Material High-flow agarose
pH Range 4 to 12
Particle Size 50μm
Matrix Ion Exchange Chromatography Media