missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto™ L Chromatography Medium

Catalog No. 45002601
Change view
Click to view available options
Quantity:
5 mL
25 mL
200 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002601 25 mL
45002603 5 mL
09928076 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45002601 Supplier Cytiva Supplier No. 17547801
Only null left

Capto L is a BioProcess chromatography medium (resin) for the capture of antibodies and antibody fragments.

Capto L is suitable for the capture of a wide range of antibody fragments such as Fabs, single-chain variable fragments (scFv), and domain antibodies (Dabs).

  • High binding capacity, low ligand leakage, and selectivity for a broad range of antibody fragments reduces process time and amount of medium.
  • Enables a platform approach for purification of antibody fragments.
  • Increased productivity, process economy, and process design flexibility due to rigid matrix.
  • The high binding capacity and selectivity for a broad range of antibody fragments makes Capto L an excellent capture platform for antibody fragments containing kappa light chains.
  • Using Capto L requires minimal optimization and it produces high purity in one separation step.
  • Capto L provides several advantages in manufacturing processes because the high selectivity of affinity capture reduces the number of subsequent polishing steps.

Specifications

Product Type Capto L chromato graphy media
Quantity 25 mL
Height (English) 7.87 in.
Height (Metric) 200mm
Flow Rate <500 cm/hr.
Ligand Type Recombinant Protein L (E. coli), mammalian free
pH Range 2 to 10
Particle Size 85μm
Support Type Highly Cross-linked Agarose
Content And Storage 2° to 8°C
For Use With (Application) High binding capacity, low ligand leakage and selectivity for a broad range of Antibody fragments reduces process time and amount of medium
Matrix Capto L Chromatography media
Show More Show Less