missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ CapSeal Bullet™ Capillary Ferrule, Short Design, 0.32 mm Column I.D.
CapSeal Bullet™™ ferrules consist of a graphite material captured in an aluminum base. This unique design keeps the ferrule from adhering to the fitting, making it easy to remove. Eliminate the headache of digging out a stuck ferrule and risking damage to your fitting
$154.02 - $546.63
Specifications
| Diameter (Metric) Inner | 0.5 mm |
|---|---|
| Description | Ferrule Type: Short |
| Material | Graphite in an Aluminum Base |
Description
Characteristics: Seals at 1/8 turn past finger tight. Reusable. A special end taper reduces graphite extrusion into fitting. Material Composition: graphite in an aluminum base. Maximum Temperature: 450°CSpecifications
| 0.5 mm | |
| Ferrule Type: Short | |
| Graphite in an Aluminum Base |
CapSeal Bullet is a registered trademark of Sigma-Aldrich Co. LLC