missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ CAPS, ULTROL™, Grade, Calbiochem™,

A zwitterionic buffer that is useful in the pH 7.9 to 11.1 range

$848.00 - $848.00

Chemical Identifiers

CAS 1135-40-6
Molecular Formula C9H19NO3S
Molecular Weight (g/mol) 221.32
MDL Number MFCD00003837
InChI Key PJWWRFATQTVXHA-UHFFFAOYSA-N
Synonym 3-(Cyclohexylamino)propanesulfonic Acid
PubChem CID 70815
IUPAC Name 3-(cyclohexylamino)propane-1-sulfonic acid
SMILES OS(=O)(=O)CCCNC1CCCCC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
2397821KG
View Documents
MilliporeSigma™
2397821KG
1 kg
Each for $848.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1135-40-6
221.32
PJWWRFATQTVXHA-UHFFFAOYSA-N
70815
OS(=O)(=O)CCCNC1CCCCC1
C9H19NO3S
MFCD00003837
3-(Cyclohexylamino)propanesulfonic Acid
3-(cyclohexylamino)propane-1-sulfonic acid

Specifications

1135-40-6
≥99.0%
MFCD00003837
Easily soluble in cold water
OS(=O)(=O)CCCNC1CCCCC1
221.32
221.3
by titration
1 kg
C9H19NO3S
3-(Cyclohexylamino)propanesulfonic Acid
PJWWRFATQTVXHA-UHFFFAOYSA-N
3-(cyclohexylamino)propane-1-sulfonic acid
70815
zwitterionic buffer
Videos
Safety and Handling

Safety and Handling

Recommended Storage : 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).

SDS
missing translation for 'documents'

missing translation for 'documents'