missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ CAPS, ULTROL™, Grade, Calbiochem™,

Catalog No. 2397821KG
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2397821KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2397821KG Supplier MilliporeSigma™ Supplier No. 2397821KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A zwitterionic buffer that is useful in the pH 7.9 to 11.1 range

Chemical Identifiers

CAS 1135-40-6
Molecular Formula C9H19NO3S
Molecular Weight (g/mol) 221.32
MDL Number MFCD00003837
InChI Key PJWWRFATQTVXHA-UHFFFAOYSA-N
Synonym 3-(Cyclohexylamino)propanesulfonic Acid
PubChem CID 70815
IUPAC Name 3-(cyclohexylamino)propane-1-sulfonic acid
SMILES OS(=O)(=O)CCCNC1CCCCC1

Specifications

Quantity 1 kg
Assay Percent Range ≥99.0%
Solubility Information Easily soluble in cold water
Formula Weight 221.3
Chemical Name or Material zwitterionic buffer
Grade by titration
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).