missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals CAPS, 0.5M buffer soln., pH 11.0

Catalog No. AAJ63138AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63138AK 250 mL
AAJ63138AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63138AK Supplier Thermo Scientific Chemicals Supplier No. J63138AK

CAS: 1135-40-6 | C9H19NO3S | 221.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1135-40-6
Molecular Formula C9H19NO3S
Molecular Weight (g/mol) 221.32
MDL Number MFCD00003837
InChI Key PJWWRFATQTVXHA-UHFFFAOYSA-N
IUPAC Name 3-(cyclohexylamino)propane-1-sulfonic acid
SMILES OS(=O)(=O)CCCNC1CCCCC1

Specifications

Concentration 0.5 M
pH 11.0
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material CAPS
Recommended Storage Ambient temperatures

RUO – Research Use Only