missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals CAPS, 0.5M buffer soln., pH 10.5

Catalog No. AAJ62446AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62446AK 250 mL
AAJ62446AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62446AK Supplier Thermo Scientific Chemicals Supplier No. J62446AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1135-40-6 | C9H19NO3S | 221.32 g/mol

CAPS (N-cyclohexyl-3-aminopropanesulfonic acid) buffer salt is used to formulate CAPS buffer, a zwitterionic buffer that is useful in the range of pH 7.9-11.1. CAPS buffer is widely used in Western and immunoblotting experiments as well as protein sequencing and identification. Used in the electrotransfer of proteins to PVDF (sc-3723) or nitrocellulose membranes (sc-3718, sc-3724). The high pH of this buffer makes it useful for the transfer of proteins with a pI > 8.5. CAPS is not one of the original Good’s buffers, although it has a similar structure to the other propanesulfonic acids and was selected as a highly water soluble buffering reagent with an optimum buffering pH of 10.4 and minimal reactivity with enzymes or proteins, minimal salt effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
CAPS (N-cyclohexyl-3-aminopropanesulfonic acid) buffer salt is used to formulate CAPS buffer, a zwitterionic buffer that is useful in the range of pH 7.9-11.1. CAPS buffer is widely used in Western and immunoblotting experiments as well as protein sequencing and identification. Used in the electrotransfer of proteins to PVDF (sc-3723) or nitrocellulose membranes (sc-3718, sc-3724). The high pH of this buffer makes it useful for the transfer of proteins with a pI>8.5. CAPS is not one of the original Good’s buffers, although it has a similar structure to the other propanesulfonic acids and was selected as a highly water soluble buffering reagent with an optimum buffering pH of 10.4 and minimal reactivity with enzymes or proteins, minimal salt effects.

Solubility
Soluble in water (100 g/L) at 20°C, and methanol (1%).

Notes
Store at room temperature. Ensure good ventilation. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1135-40-6
Molecular Formula C9H19NO3S
Molecular Weight (g/mol) 221.32
MDL Number MFCD00003837
InChI Key PJWWRFATQTVXHA-UHFFFAOYSA-N
IUPAC Name 3-(cyclohexylamino)propane-1-sulfonic acid
SMILES OS(=O)(=O)CCCNC1CCCCC1

Specifications

Concentration 0.5 M
pH 10.5
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material CAPS
Recommended Storage Ambient temperatures

RUO – Research Use Only