missing translation for 'onlineSavingsMsg'
Learn More

Capreomycin sulfate, 90%

CAS: 1405-37-4 | C25H46N14O12S | 766.79 g/mol

$268.33 - $268.33

Chemical Identifiers

CAS 1405-37-4
Molecular Formula C25H46N14O12S
Molecular Weight (g/mol) 766.79
MDL Number MFCD00079032
InChI Key AJQVUIHGEOLMDY-SOCRLDLMNA-N
IUPAC Name 3,6-diamino-N-{[(8Z)-15-amino-11-(2-amino-3,4,5,6-tetrahydropyrimidin-4-yl)-8-[(carbamoylamino)methylidene]-2-(hydroxymethyl)-3,6,9,12,16-pentaoxo-1,4,7,10,13-pentaazacyclohexadecan-5-yl]methyl}hexanamide; sulfuric acid
SMILES OS(O)(=O)=O.NCCCC(N)CC(=O)NCC1NC(=O)C(CO)NC(=O)C(N)CNC(=O)C(NC(=O)\C(NC1=O)=C\NC(N)=O)C1CCN=C(N)N1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6668403
View Documents
Thermo Scientific Chemicals
J6668403
1 g
Each for $268.33
Only null left
 
Description

Description

Thought to inhibit protein synthesis by inhibiting normal ribosomal functions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Thought to inhibit protein synthesis by inhibiting normal ribosomal functions
Specifications

Chemical Identifiers

1405-37-4
766.79
AJQVUIHGEOLMDY-SOCRLDLMNA-N
OS(O)(=O)=O.NCCCC(N)CC(=O)NCC1NC(=O)C(CO)NC(=O)C(N)CNC(=O)C(NC(=O)\C(NC1=O)=C\NC(N)=O)C1CCN=C(N)N1
C25H46N14O12S
MFCD00079032
3,6-diamino-N-{[(8Z)-15-amino-11-(2-amino-3,4,5,6-tetrahydropyrimidin-4-yl)-8-[(carbamoylamino)methylidene]-2-(hydroxymethyl)-3,6,9,12,16-pentaoxo-1,4,7,10,13-pentaazacyclohexadecan-5-yl]methyl}hexanamide; sulfuric acid

Specifications

Capreomycin Sulfate
1405-37-4
1 g
C25H46N14O12S
1377°C
AJQVUIHGEOLMDY-SOCRLDLMNA-N
3,6-diamino-N-{[(8Z)-15-amino-11-(2-amino-3,4,5,6-tetrahydropyrimidin-4-yl)-8-[(carbamoylamino)methylidene]-2-(hydroxymethyl)-3,6,9,12,16-pentaoxo-1,4,7,10,13-pentaazacyclohexadecan-5-yl]methyl}hexanamide; sulfuric acid
766.78
Capreomycin sulfate
Powder
White
>110°C (230°F)
MFCD00079032
Freely soluble in water
OS(O)(=O)=O.NCCCC(N)CC(=O)NCC1NC(=O)C(CO)NC(=O)C(N)CNC(=O)C(NC(=O)\C(NC1=O)=C\NC(N)=O)C1CCN=C(N)N1
766.79
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H302-H312-H332
May damage fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c

H302+H312+H332-H360

EINECSNumber : 215-776-8

RTECSNumber : EX8930000

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only