missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Cannula
GREENER_CHOICE

Catalog No. 31502002
Change view
Click to view available options
Needle Gauge:
16 G
18 G
20 G
22 G
Length (Metric) Needle:
460 mm
600 mm
900 mm
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Length (Metric) Needle
31502002 22 G 900 mm
31501991 16 G 460 mm
31501992 16 G 600 mm
31501993 16 G 900 mm
31501994 18 G 460 mm
31501995 18 G 600 mm
31501996 18 G 900 mm
31501997 20 G 460 mm
31501998 20 G 600 mm
31501999 20 G 900 mm
31502000 22 G 460 mm
31502001 22 G 600 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 31502002 Supplier Synthware™ Supplier No. C702236

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cannula for the transfer and handling of air sensitive materials.

Specifications

Material Stainless Steel
Color Silver
Needle Gauge 22 G
For Use With (Application) Organic Synthesis
Length (English) Needle 35.4 in.
Product Type Cannula
Quantity 1 Ea.
Length (Metric) Needle 900 mm
Graduated No
Sterility Non-sterile