missing translation for 'onlineSavingsMsg'
Learn More

Canertinib dihydrochloride, Thermo Scientific™

Catalog No. p-7238578
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468932500 250 mg
AC468930010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468932500 Supplier Thermo Scientific Chemicals Supplier No. 468932500
Only null left

Chemical Identifiers

CAS 289499-45-2
Molecular Formula C24 H25 Cl F N5 O3 · 2 H Cl

Specifications

Quantity 250 mg
Formula Weight 558.87
MDL Number MFCD09954112
Physical Form Powder
Color White to Beige
Purity ≥97.5%
InChI Key JZZFDCXSFTVOJY-UHFFFAOYSA-N
SMILES Cl.Cl.FC1=CC=C(NC2=C3C=C(NC(=O)C=C)C(OCCCN4CCOCC4)=CC3=NC=N2)C=C1Cl
IUPAC Name N-{4-[(3-chloro-4-fluorophenyl)amino]-7-[3-(morpholin-4-yl)propoxy]quinazolin-6-yl}prop-2-enamide dihydrochloride
Molecular Weight (g/mol) 558.86
Chemical Name or Material Canertinib dihydrochloride
Show More Show Less
Hazard Category Exclamation mark
Recommended Storage Normal conditions