missing translation for 'onlineSavingsMsg'
Learn More

Canertinib dihydrochloride, Thermo Scientific™

Catalog No. AC468932500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468932500 250 mg
AC468930010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468932500 Supplier Thermo Scientific Chemicals Supplier No. 468932500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 289499-45-2
Molecular Formula C24 H25 Cl F N5 O3 · 2 H Cl

Specifications

Quantity 250 mg
Formula Weight 558.87
Physical Form Powder
Color White to Beige
Purity ≥97.5%
Chemical Name or Material Canertinib dihydrochloride
Hazard Category Exclamation mark
Recommended Storage Normal conditions