missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canertinib dihydrochloride, Thermo Scientific™
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 558.87 |
| Physical Form | Powder |
| Color | White to Beige |
| Purity | ≥97.5% |
| Chemical Name or Material | Canertinib dihydrochloride |
Safety and Handling
| Hazard Category | Exclamation mark |
| Recommended Storage | Normal conditions |