missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-Camphor 95.0+%, TCI America™
$32.28 - $174.67
Chemical Identifiers
| CAS | 76-22-2 |
|---|---|
| Molecular Formula | C10H16O |
| Molecular Weight (g/mol) | 152.24 |
| MDL Number | MFCD00074738,MFCD00064149 |
| InChI Key | DSSYKIVIOFKYAU-UHFFFAOYNA-N |
| Synonym | camphor, dl-camphor, 2-camphanone, 2-bornanone, l---camphor, +/--camphor, root bark oil, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, bornan-2-one, alphanon |
| PubChem CID | 2537 |
| ChEBI | CHEBI:36773 |
| IUPAC Name | 1,7,7-trimethylbicyclo[2.2.1]heptan-2-one |
| SMILES | CC1(C)C2CCC1(C)C(=O)C2 |
Chemical Identifiers
| C10H16O | |
| MFCD00074738,MFCD00064149 | |
| camphor, dl-camphor, 2-camphanone, 2-bornanone, l---camphor, +/--camphor, root bark oil, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, bornan-2-one, alphanon | |
| CHEBI:36773 | |
| CC1(C)C2CCC1(C)C(=O)C2 |
Specifications
| 76-22-2 | |
| White | |
| MFCD00074738,MFCD00064149 | |
| camphor, dl-camphor, 2-camphanone, 2-bornanone, l---camphor, +/--camphor, root bark oil, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, bornan-2-one, alphanon | |
| CC1(C)C2CCC1(C)C(=O)C2 | |
| 152.24 | |
| CHEBI:36773 | |
| ≥95.0% (GC) | |
| (+/-)-Camphor |
| 174°C | |
| C10H16O | |
| 2717 | |
| DSSYKIVIOFKYAU-UHFFFAOYNA-N | |
| 1,7,7-trimethylbicyclo[2.2.1]heptan-2-one | |
| 2537 | |
| 152.24 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0308&(4)-0601
RTECSNumber : EX1234000
TSCA : Yes