accessibility menu, dialog, popup

UserName

H3CH3CCH3O

CAS RN 76-22-2

IUPAC Name: 1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
Synonyms: camphor gum camphor laurel camphor formosa camphor kampfer spirit of camphor root bark oil japan camphor 2-camphanone
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: DSSYKIVIOFKYAU-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1(C)C(=O)C2
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (7)
Grade 
  • (1)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (5)
  • (1)
  • (5)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (7)
  • (7)
  • (1)
  • (5)
  • (2)
  • (1)