missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Camphor 95.0+%, TCI America™

Catalog No. C001125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C001125G 25 g
C0011500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C001125G Supplier TCI America Supplier No. C001125G

Chemical Identifiers

CAS 76-22-2
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00074738,MFCD00064149
InChI Key DSSYKIVIOFKYAU-UHFFFAOYNA-N
Synonym camphor, dl-camphor, 2-camphanone, 2-bornanone, l---camphor, +/--camphor, root bark oil, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, bornan-2-one, alphanon
PubChem CID 2537
ChEBI CHEBI:36773
IUPAC Name 1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
SMILES CC1(C)C2CCC1(C)C(=O)C2

Specifications

Melting Point 174°C
Color White
Quantity 25 g
UN Number 2717
Formula Weight 152.24
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-Camphor
EINECSNumber (4)-0308&(4)-0601
RTECSNumber EX1234000
TSCA Yes